Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172804.2 | NP_766392.2 | 5 | 838 | ||
GenBank mRNA | AB102950.1 | - | 4 | 1170 | ||
AK032606.1 | - | 6 | 1617 | |||
AK137129.1 | - | 5 | 846 | |||
BC056932.1 | - | 4 | 948 | 5708126 |