Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172708.3 | NP_766296.1 | 6 | 1063 | ||
GenBank mRNA | AB220919.1 | - | 7 | 1109 | ||
AK044445.1 | - | 6 | 1064 | |||
AK170454.1 | - | 5 | 800 | |||
BC089590.1 | - | 7 | 1182 | 6759981 |