Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115010.1 | NP_001108482.1 | 8 | 1823 | ||
NM_172714.4 | NP_766302.2 | 8 | 1691 | |||
GenBank mRNA | AK029288.1 | - | 9 | 1137 | ||
AK030894.1 | - | 8 | 1825 | |||
AK134320.1 | - | 8 | 1689 | |||
AK220225.1 | - | 8 | 1686 | |||
BC106126.1 | - | 7 | 1327 | |||
BC125580.1 | - | 6 | 818 | |||
BC145025.1 | - | 6 | 818 | 9053573 |