Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMK2B SIRNA Assays ›
Gene Symbol:
CAMK2B
Gene Name:
calcium/calmodulin dependent protein kinase II beta
Gene Aliases:
CAM2, CAMK2, CAMKB
Chromosome Location:
Chr.7: 44217150 - 44325994 on Build GRCh38
UniGene ID:
Hs.351887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
9477
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMK2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220.4
NP_001211.3
7, 8
725
NM_001293170.1
NP_001280099.1
7, 8
725
NM_172078.2
NP_742075.1
7, 8
725
NM_172079.2
NP_742076.1
7, 8
725
NM_172080.2
NP_742077.1
7, 8
725
NM_172081.2
NP_742078.1
7, 8
725
NM_172082.2
NP_742079.1
7, 8
725
NM_172083.2
NP_742080.1
7, 8
725
NM_172084.2
NP_742081.1
7, 8
725
XM_005249862.3
XP_005249919.1
7, 8
601
XM_005249864.3
XP_005249921.1
7, 8
601
XM_006715781.2
XP_006715844.1
7, 8
601
XM_006715784.2
XP_006715847.1
7, 8
601
XM_011515547.1
XP_011513849.1
7, 8
601
XM_011515549.1
XP_011513851.1
7, 8
601
XM_011515550.1
XP_011513852.1
7, 8
601
XM_011515551.1
XP_011513853.1
7, 8
601
XM_011515552.1
XP_011513854.1
7, 8
601
XM_011515553.1
XP_011513855.1
7, 8
601
XM_011515554.1
XP_011513856.1
7, 8
601
XM_011515555.1
XP_011513857.1
7, 8
601
XM_011515556.1
XP_011513858.1
7, 8
601
XM_011515557.1
XP_011513859.1
7, 8
601
XM_011515558.2
XP_011513860.1
7, 8
601
XM_011515559.1
XP_011513861.1
7, 8
510
XM_017012660.1
XP_016868149.1
7, 8
601
XM_017012661.1
XP_016868150.1
7, 8
601
XM_017012662.1
XP_016868151.1
7, 8
601
XM_017012663.1
XP_016868152.1
7, 8
510
XM_017012664.1
XP_016868153.1
7, 8
601
XM_017012665.1
XP_016868154.1
7, 8
601
XM_017012666.1
XP_016868155.1
7, 8
510
XM_017012667.1
XP_016868156.1
7, 8
510
XM_017012668.1
XP_016868157.1
7, 8
510
XM_017012669.1
XP_016868158.1
7, 8
510
XM_017012670.1
XP_016868159.1
7, 8
510
GenBank mRNA
AA399393.1
-
3, 4
222
AF078803.1
-
7, 8
518
AF081572.1
-
7, 8
518
AF081924.1
-
7, 8
518
AF083419.1
-
7, 8
518
AF112471.1
-
7, 8
561
AF112472.1
-
7, 8
561
AF140350.1
-
7, 8
518
AJ252236.1
-
7, 8
602
AJ252237.1
-
7, 8
525
AJ252238.1
-
7, 8
515
AK222703.1
-
7, 8
549
AK290148.1
-
7, 8
590
AK294138.1
-
5, 6
448
AK315663.1
-
7, 8
632
BC019070.2
-
7, 8
719
5014712
BX445032.2
-
7, 8
534
DA069361.1
-
6, 7
377
DB175099.1
-
7, 8
503
U23460.1
-
7, 8
590
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein phosphorylation
signal transduction
positive regulation of neuron projection development
regulation of skeletal muscle adaptation
positive regulation of phospholipase A2 activity
positive regulation of GTPase activity
protein autophosphorylation
regulation of long-term neuronal synaptic plasticity
cell projection morphogenesis
regulation of synapse structural plasticity
regulation of calcium ion transport
interferon-gamma-mediated signaling pathway
regulation of dendritic spine development
positive regulation of dendritic spine morphogenesis
positive regulation of synapse maturation
regulation of cellular response to heat
actin binding
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
Ras guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
protein kinase binding
protein homodimerization activity
phospholipase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-kprhh:80/10.239.160.57:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline