Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap5z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172725.2 | NP_766313.2 | 12 | 1650 | ||
GenBank mRNA | AK040924.1 | - | 10 | 1202 | ||
AK082759.1 | - | 4 | 477 | |||
AK129135.1 | - | 9 | 2848 | |||
AK146431.1 | - | 12 | 1528 | |||
AK152402.1 | - | 12 | 1528 | |||
AK153787.1 | - | 11 | 3169 | |||
AK184998.1 | - | 2 | 53 | |||
AK196939.1 | - | 2 | 53 |