Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E130309D02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172726.4 | NP_766314.3 | 2 | 484 | ||
GenBank mRNA | AK041667.1 | - | 2 | 430 | ||
AK050733.1 | - | 2 | 398 | |||
AK053801.1 | - | 2 | 485 | |||
AK054218.1 | - | 2 | 185 | |||
AK086571.1 | - | 3 | 1029 | |||
AK087531.1 | - | 2 | 485 | |||
AK144964.1 | - | 2 | 465 | |||
AK172614.1 | - | 2 | 431 | |||
BC058940.1 | - | 2 | 262 | |||
BC079853.1 | - | 2 | 418 | 5716912 | ||
BC082321.1 | - | 2 | 228 | 6411746 |