Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172730.2 | NP_766318.2 | 5, 6 | 583 | ||
GenBank mRNA | AK039818.1 | - | 5, 6 | 562 | ||
AK046253.1 | - | 1, 2 | 2428 | |||
AK081505.1 | - | 4, 5 | 3228 | |||
AK156259.1 | - | 5, 6 | 566 | |||
BC016607.1 | - | 3, 4 | 351 | |||
BC051147.1 | - | 5, 6 | 577 | 6595828 |