Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172740.2 | NP_766328.2 | 4 | 1382 | ||
GenBank mRNA | AK045435.1 | - | 2 | 1040 | ||
AK045821.1 | - | 4 | 1295 | |||
BC050249.1 | - | 3 | 1267 | |||
BC055817.1 | - | 4 | 1302 | 3595825 |