Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172757.3 | NP_766345.3 | 5 | 693 | ||
GenBank mRNA | AK047782.1 | - | 5 | 724 | ||
AK052635.1 | - | 5 | 720 | |||
AK136424.1 | - | 5 | 703 | |||
AK140522.1 | - | 5 | 711 | |||
AK163715.1 | - | 5 | 680 | |||
AK165541.1 | - | 5 | 587 | |||
BC030373.1 | - | 1 | 38 | |||
BC065060.1 | - | 5 | 693 | 6402217 |