Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172760.3 | NP_766348.3 | 6, 7 | 668 | ||
GenBank mRNA | AK037067.1 | - | 6, 7 | 661 | ||
AK157875.1 | - | 606 | ||||
BC018516.1 | - | 4, 5 | 557 | 3155913 | ||
BC026617.1 | - | 1, 2 | 613 | |||
BC058752.1 | - | 6, 7 | 651 | 6485393 | ||
BC138887.1 | - | 6, 7 | 625 | 8861909 |