Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfrkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172766.3 | NP_766354.2 | 17 | 1937 | ||
GenBank mRNA | AK041206.1 | - | 17 | 2175 | ||
AK049804.1 | - | 3 | 312 | |||
BC033595.1 | - | 17 | 1927 | 4947357 | ||
BC059861.1 | - | 17 | 1984 | |||
BC059900.1 | - | 8 | 1584 | |||
BC060076.1 | - | 17 | 2166 |