Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145957.1 | NP_001139429.1 | 3 | 287 | ||
NM_172767.3 | NP_766355.3 | 4 | 363 | |||
GenBank mRNA | AK019914.1 | - | 4 | 317 | ||
AK041397.1 | - | 4 | 330 | |||
AK053665.1 | - | 4 | 307 | |||
AK054497.1 | - | 4 | 336 | |||
AK136934.1 | - | 4 | 336 | |||
AK161193.1 | - | 3 | 274 | |||
AY366502.1 | - | 4 | 333 | |||
BC065153.1 | - | 3 | 217 | 6418714 |