Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172790.2 | NP_766378.1 | 8 | 920 | ||
GenBank mRNA | AK032429.1 | - | 8 | 923 | ||
AK035533.1 | - | 8 | 908 | |||
AK090124.1 | - | 8 | 920 | |||
AK144017.1 | - | 8 | 930 | |||
BC117907.1 | - | 8 | 871 | 40097189 | ||
BC117908.1 | - | 8 | 871 | 40097194 |