Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172796.2 | NP_766384.2 | 2 | 399 | ||
GenBank mRNA | AK036579.1 | - | 2 | 400 | ||
AK050355.1 | - | 2 | 353 | |||
AY261800.1 | - | 2 | 307 | |||
AY261801.1 | - | 2 | 289 |