Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172710.3 | NP_766298.2 | 8 | 1534 | ||
GenBank mRNA | AK030487.1 | - | 8 | 1399 | ||
AK037220.1 | - | 8 | 1398 | |||
AK122362.1 | - | 8 | 1534 | |||
BC094607.1 | - | 7 | 1079 | 6837048 | ||
BC117915.1 | - | 7 | 1035 | 40097358 | ||
BC117916.1 | - | 7 | 1037 | 40097359 |