Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115010.1 | NP_001108482.1 | 13 | 3280 | ||
NM_172714.4 | NP_766302.2 | 13 | 3148 | |||
GenBank mRNA | AK029288.1 | - | 14 | 2594 | ||
AK030894.1 | - | 13 | 3281 | |||
AK220225.1 | - | 13 | 3143 |