Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172721.2 | NP_766309.2 | 11 | 1909 | ||
GenBank mRNA | AK031815.1 | - | 11 | 1903 | ||
AK034447.1 | - | 11 | 1837 | |||
AK053794.1 | - | 11 | 1910 | |||
AK134291.1 | - | 11 | 1917 | |||
AK163976.1 | - | 3 | 458 | |||
BC009095.1 | - | 11 | 1862 | 3158218 | ||
BC024091.1 | - | 5 | 785 |