Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276287.1 | NP_001263216.1 | 14 | 3240 | ||
NM_181066.3 | NP_851411.1 | 14 | 3142 | |||
GenBank mRNA | AF226663.1 | - | 4 | 1689 | ||
AK035518.1 | - | 1 | 1406 | |||
AK084987.1 | - | 13 | 3713 | |||
AK085091.1 | - | 14 | 3131 | |||
AK170269.1 | - | 14 | 3137 | |||
BC068301.1 | - | 14 | 3206 | 30061848 |