Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001246.3 | NP_001237.1 | 2 | 248 | ||
NM_203468.2 | NP_982293.1 | 2 | 248 | |||
XM_011519212.2 | XP_011517514.1 | 1 | 65 | |||
GenBank mRNA | AF144748.1 | - | 2 | 194 | ||
BC035738.1 | - | 2 | 242 | 5730586 | ||
BQ883059.1 | - | 272 | ||||
CB045339.1 | - | 2 | 273 | |||
EF495152.1 | - | 2 | 194 | |||
U91510.1 | - | 2 | 215 |