Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dera | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172733.1 | NP_766321.1 | 9 | 1210 | ||
GenBank mRNA | AK051983.1 | - | 1 | 1835 | ||
AK075963.1 | - | 9 | 1235 | |||
AK083298.1 | - | 9 | 1210 | |||
AK145820.1 | - | 9 | 1246 | |||
AK150273.1 | - | 9 | 1236 | |||
AK150712.1 | - | 9 | 1206 | |||
AK152462.1 | - | 9 | 1214 | |||
AK168204.1 | - | 9 | 1246 | |||
AK169418.1 | - | 9 | 1230 | |||
BC016218.1 | - | 9 | 1217 | 4484163 |