Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tufm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172745.3 | NP_766333.1 | 8, 9 | 1166 | ||
GenBank mRNA | AK075857.1 | - | 8, 9 | 1090 | ||
AK084724.1 | - | 8, 9 | 1147 | |||
AK152858.1 | - | 8, 9 | 1161 | |||
AK153135.1 | - | 8, 9 | 1161 | |||
BC100596.1 | - | 8, 9 | 1159 | 6762419 |