Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145957.1 | NP_001139429.1 | 9 | 1217 | ||
NM_172767.3 | NP_766355.3 | 10 | 1293 | |||
GenBank mRNA | AK019914.1 | - | 10 | 1247 | ||
AK041397.1 | - | 10 | 1260 | |||
AK053665.1 | - | 10 | 1237 | |||
AK054497.1 | - | 10 | 1266 | |||
AK136934.1 | - | 10 | 1266 | |||
AK161193.1 | - | 9 | 1204 | |||
AK193979.1 | - | 1 | 64 | |||
AY366502.1 | - | 10 | 1263 | |||
BC004727.1 | - | 4 | 381 | 3582797 | ||
BC065153.1 | - | 9 | 1147 | 6418714 |