Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040059.1 | NP_001035148.1 | 2 | 272 | ||
NM_001251.2 | NP_001242.2 | 2 | 354 | |||
GenBank mRNA | AK222492.1 | - | 2 | 201 | ||
AK222514.1 | - | 2 | 200 | |||
AK290238.1 | - | 2 | 214 | |||
AK298055.1 | - | 2 | 203 | |||
AK301223.1 | - | 2 | 106 | |||
BC015557.2 | - | 2 | 226 | 3050731 | ||
BT009923.1 | - | 2 | 163 | |||
BX438711.2 | - | 2 | 111 | |||
BX647524.1 | - | 2 | 121 | |||
DB187273.1 | - | 2 | 374 | |||
S57235.1 | - | 2 | 178 |