Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161430.2 | NP_001154902.1 | 5 | 1248 | ||
NM_001290532.1 | NP_001277461.1 | 5 | 1333 | |||
NM_001290533.1 | NP_001277462.1 | 4 | 1339 | |||
NM_172782.4 | NP_766370.2 | 4 | 1285 | |||
GenBank mRNA | AK032102.1 | - | 4 | 1256 | ||
AK144349.1 | - | 4 | 1271 | |||
BC064727.1 | - | 4 | 1260 | |||
BC068166.1 | - | 4 | 1238 | 30361776 | ||
BC094570.1 | - | 4 | 1202 | 30361291 | ||
BC118503.1 | - | 4 | 1213 | 40106658 |