Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3mbtl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172787.2 | NP_766375.1 | 21, 22 | 2592 | ||
GenBank mRNA | AK030412.1 | - | 8, 9 | 1190 | ||
AK045667.1 | - | 21, 22 | 2593 | |||
AK129443.1 | - | 17, 18 | 2137 | |||
BC082309.1 | - | 20, 21 | 2462 | 30356672 | ||
BC085192.1 | - | 20, 21 | 2697 | 30694487 |