Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172790.2 | NP_766378.1 | 28 | 4157 | ||
GenBank mRNA | AK090124.1 | - | 28 | 4157 |