Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172838.3 | NP_766426.1 | 3 | 348 | ||
GenBank mRNA | AK028284.1 | - | 3 | 346 | ||
AK029076.1 | - | 3 | 348 | |||
BC111902.1 | - | 2 | 301 | 40062303 | ||
BC113803.1 | - | 2 | 301 | 40062304 |