Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEN1-CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037709.1 | 6 | 2065 | |||
GenBank mRNA | AK097104.1 | - | 6 | 2022 | ||
BX647274.1 | - | 6 | 2051 |
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258.2 | NP_001249.1 | 4 | 305 | ||
GenBank mRNA | DB455174.1 | - | 4 | 306 | ||
X66357.1 | - | 3 | 317 |