Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172881.3 | NP_766469.1 | 1 | 115 | ||
GenBank mRNA | AK083294.1 | - | 1 | 115 | ||
AK149501.1 | - | 1 | 102 | |||
BC113789.1 | - | 1 | 105 | 40061872 |