Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933405O20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172901.2 | NP_766489.1 | 1 | 110 | ||
GenBank mRNA | AK077105.1 | - | 1 | 111 | ||
BC060958.1 | - | 1 | 87 | 6744040 |