Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001096.2 | NP_001087.2 | 4, 5 | 516 | ||
NM_001303274.1 | NP_001290203.1 | 4, 5 | 523 | |||
NM_001303275.1 | NP_001290204.1 | 4, 5 | 523 | |||
NM_198830.1 | NP_942127.1 | 4, 5 | 516 | |||
XM_005257395.1 | XP_005257452.1 | 4, 5 | 565 | |||
XM_017024688.1 | XP_016880177.1 | 4, 5 | 565 | |||
GenBank mRNA | AB210035.1 | - | 4, 5 | 463 | ||
AK295675.1 | - | 4, 5 | 523 | |||
AK312315.1 | - | 4, 5 | 520 | |||
BC006195.2 | - | 4, 5 | 484 | 2959339 | ||
BG037168.1 | - | 536 | ||||
DB072009.1 | - | 4, 5 | 520 | |||
U18197.1 | - | 4, 5 | 424 | |||
X64330.1 | - | 4, 5 | 437 |