Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172861.3 | NP_766449.1 | 2, 3 | 616 | ||
GenBank mRNA | AK044448.1 | - | 2, 3 | 609 | ||
AK163358.1 | - | 2, 3 | 660 | |||
AK173217.1 | - | 2, 3 | 592 | |||
BC061928.1 | - | 2, 3 | 592 | 5359765 |