Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172863.4 | NP_766451.3 | 2 | 326 | ||
GenBank mRNA | AK034880.1 | - | 2 | 327 | ||
BC092534.1 | - | 2 | 259 | 30249621 | ||
BQ890991.1 | - | 2 | 245 |