Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172866.3 | NP_766454.1 | 7 | 1038 | ||
GenBank mRNA | AK089288.1 | - | 7 | 821 | ||
AK129103.1 | - | 7 | 824 | |||
AK170117.1 | - | 7 | 835 | |||
BC086614.1 | - | 7 | 844 | 30533256 |