Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172870.4 | NP_766458.3 | 4 | 557 | ||
GenBank mRNA | AK030323.1 | - | 4 | 557 | ||
AK031440.1 | - | 6 | 758 | |||
AY438375.1 | - | 4 | 532 | |||
BC110483.2 | - | 3 | 437 | 40048720 | ||
BC110484.1 | - | 3 | 437 | 40048722 |