Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Azin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047522.1 | - | 4, 5 | 640 | ||
AK077201.1 | - | 4, 5 | 640 | |||
AK148717.1 | - | 4, 5 | 644 | |||
BC138973.1 | - | 4, 5 | 552 | 8861995 | ||
BC138974.1 | - | 4, 5 | 552 | 8861996 |