Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pramef8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172877.2 | NP_766465.1 | 3, 4 | 1041 | ||
GenBank mRNA | AK029142.1 | - | 3, 4 | 1042 | ||
BC056931.1 | - | 2, 3 | 415 | 6816019 |