Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kirrel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172898.3 | NP_766486.1 | 5 | 1205 | ||
GenBank mRNA | AK049284.1 | - | 5 | 1206 | ||
AK083069.1 | - | 5 | 1347 | |||
AY305303.1 | - | 4 | 343 | |||
BC052773.1 | - | 5 | 668 | 6314924 | ||
BC158021.1 | - | 5 | 713 | 9007327 |