Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172807.4 | NP_766395.2 | 5 | 974 | ||
GenBank mRNA | AK028532.1 | - | 5 | 954 | ||
AK043316.1 | - | 5 | 975 | |||
BC037196.1 | - | 1 | 109 | |||
BC119282.1 | - | 5 | 952 | 8734031 | ||
BC119310.1 | - | 5 | 952 | 8734059 |