Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109.4 | NP_001100.3 | 14 | 1504 | ||
NM_001164489.1 | NP_001157961.1 | 14 | 1504 | |||
NM_001164490.1 | NP_001157962.1 | 13 | 1387 | |||
XM_011539117.2 | XP_011537419.2 | 15 | 1685 | |||
XM_017015465.1 | XP_016870954.1 | 15 | 1685 | |||
XM_017015466.1 | XP_016870955.1 | 15 | 1687 | |||
GenBank mRNA | AK301147.1 | - | 13 | 1352 | ||
AK312834.1 | - | 14 | 1469 | |||
BC064500.1 | - | 14 | 1451 | |||
BC115404.1 | - | 14 | 1423 | 40075505 | ||
D26579.1 | - | 14 | 1427 | |||
DA936506.1 | - | 5 | 551 |