Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nyap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172849.3 | NP_766437.2 | 4 | 1504 | ||
GenBank mRNA | AB429290.1 | - | 4 | 899 | ||
AK034747.1 | - | 5 | 1519 | |||
AK043512.1 | - | 4 | 1143 | |||
AK049778.1 | - | 5 | 1417 | |||
AK129372.1 | - | 1 | 143 | |||
AK137094.1 | - | 4 | 1405 | |||
BC082310.1 | - | 4 | 1504 | 30355797 |