Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172872.3 | NP_766460.2 | 10 | 3245 | ||
GenBank mRNA | AK081525.1 | - | 10 | 3183 | ||
BC060737.1 | - | 10 | 3259 | 4191502 |