Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172892.3 | NP_766480.1 | 16 | 3189 | ||
GenBank mRNA | AK036354.1 | - | 16 | 3191 | ||
AK038937.1 | - | 16 | 3194 | |||
BC089161.1 | - | 16 | 3178 | 30440173 |