Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175256.5 | NP_780465.4 | 2 | 435 | ||
GenBank mRNA | AK028518.1 | - | 2 | 436 | ||
AK163128.1 | - | 2 | 436 | |||
BB639887.1 | - | 2 | 436 |