Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077707.1 | NP_001071175.1 | 2 | 449 | ||
NM_001284354.1 | NP_001271283.1 | 2 | 449 | |||
NM_172937.3 | NP_766525.3 | 2 | 449 | |||
GenBank mRNA | AK011809.1 | - | 2 | 374 | ||
AK048265.1 | - | 2 | 407 | |||
AK053448.1 | - | 2 | 450 | |||
AK082160.1 | - | 2 | 453 | |||
AK085285.1 | - | 2 | 387 | |||
AY162264.1 | - | 2 | 355 | |||
AY162266.1 | - | 2 | 355 | |||
BC055003.1 | - | 2 | 420 | 6468603 |