Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172938.3 | NP_766526.3 | 3, 4 | 637 | ||
GenBank mRNA | AF397911.1 | - | 3, 4 | 505 | ||
AK034171.1 | - | 3, 4 | 476 | |||
AK220309.1 | - | 1, 2 | 21 | |||
BC043310.1 | - | 3, 4 | 432 | 5067233 |