Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gphn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145965.2 | NP_666077.2 | 3, 4 | 332 | ||
NM_172952.3 | NP_766540.2 | 3, 4 | 332 | |||
GenBank mRNA | AK082353.1 | - | 3, 4 | 254 | ||
AK144569.1 | - | 3, 4 | 335 | |||
BC127058.1 | - | 3, 4 | 254 | 40109771 |