Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc28a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172980.2 | NP_766568.1 | 4 | 1421 | ||
GenBank mRNA | AB485921.1 | - | 4 | 720 | ||
AK080957.1 | - | 4 | 1422 | |||
AK152776.1 | - | 3 | 356 | |||
BC055376.1 | - | 3 | 315 | 30077988 |