Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050894.1 | - | 2, 3 | 230 | ||
BC050089.1 | - | 2, 3 | 228 | 30018732 |