Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zxdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030260.3 | NP_084536.2 | 1 | 968 | ||
NM_173002.3 | NP_766590.1 | 1 | 968 | |||
GenBank mRNA | AK030287.1 | - | 1 | 884 | ||
AK042190.1 | - | 1 | 895 | |||
AK042392.1 | - | 1 | 878 | |||
AK044429.1 | - | 1 | 861 | |||
AK044529.1 | - | 1 | 892 | |||
AK044948.1 | - | 1 | 862 | |||
AK047079.1 | - | 1 | 852 | |||
AK047538.1 | - | 1 | 884 | |||
AK080307.1 | - | 1 | 862 | |||
AK081005.1 | - | 1 | 970 | |||
AK141613.1 | - | 1 | 845 | |||
AK169517.1 | - | 1 | 855 | |||
BC003332.1 | - | 1 | 952 | 3256958 |